Order Entry
Netherlands
ContactUsLinkComponent
1-[2-(Trifluoromethyl)-3-pyridyl]cyclopropanecarbonitrile
1-[2-(Trifluoromethyl)-3-pyridyl]cyclopropanecarbonitrile
Catalog # APOSPC520373-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[2-(Trifluoromethyl)-3-pyridyl]cyclopropanecarbonitrile
Catalog # APOSPC520373-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :212.171 g/mol
Storage Temperature:Ambient
CAS Number:1427012-75-6
Formula:C₁₀H₇F₃N₂
MDL Number:MFCD23134897