Methyl 5-bromo-2-(difluoromethyl)benzoate
Catalog # APOSPC520315-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:265.051 g/mol
Storage Temperature:Ambient
CAS Number:1630983-08-2
Formula:C₉H₇BrF₂O₂
MDL Number:MFCD30496628