Order Entry
Netherlands
ContactUsLinkComponent
4-(Difluoromethyl)-3,5-dimethoxy-phenol
4-(Difluoromethyl)-3,5-dimethoxy-phenol
Catalog # APOSPC520300-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Difluoromethyl)-3,5-dimethoxy-phenol
Catalog # APOSPC520300-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :204.171 g/mol
Storage Temperature:Ambient
CAS Number:2092380-84-0
Formula:C₉H₁₀F₂O₃
MDL Number:MFCD30496619