[2-(Difluoromethyl)benzothiophen-5-yl]azinate
Catalog # APOSPC520238-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:229.203 g/mol
Storage Temperature:Ambient
Formula:C₉H₅F₂NO₂S
MDL Number:MFCD30496608