4-[5-(Difluoromethyl)-2-thienyl]morpholine
Catalog # APOSPC520210-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.252 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₁F₂NOS
MDL Number:MFCD29767690