Order Entry
Netherlands
ContactUsLinkComponent
6-Bromo-3-(difluoromethyl)-7-methyl-imidazo[1,2-a]pyridine
6-Bromo-3-(difluoromethyl)-7-methyl-imidazo[1,2-a]pyridine
Catalog # APOSPC520152-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Bromo-3-(difluoromethyl)-7-methyl-imidazo[1,2-a]pyridine
Catalog # APOSPC520152-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :261.066 g/mol
Storage Temperature:Ambient
CAS Number:2092048-68-3
Formula:C₉H₇BrF₂N₂
MDL Number:MFCD29767633