Order Entry
Netherlands
ContactUsLinkComponent
4,6-Dichloro-3-(difluoromethyl)-1H-indole
4,6-Dichloro-3-(difluoromethyl)-1H-indole
Catalog # APOSPC520109-1G
Supplier:  Apollo Scientific
CAS Number:  
4,6-Dichloro-3-(difluoromethyl)-1H-indole
Catalog # APOSPC520109-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :236.046 g/mol
Storage Temperature:Ambient
Formula:C₉H₅Cl₂F₂N
MDL Number:MFCD29767592