Order Entry
Netherlands
ContactUsLinkComponent
6-Bromo-3-(difluoromethyl)-N,N-dimethyl-1H-indol-2-amine
6-Bromo-3-(difluoromethyl)-N,N-dimethyl-1H-indol-2-amine
Catalog # APOSPC520090-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Bromo-3-(difluoromethyl)-N,N-dimethyl-1H-indol-2-amine
Catalog # APOSPC520090-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :289.119 g/mol
Storage Temperature:Ambient
CAS Number:2091011-15-1
Formula:C₁₁H₁₁BrF₂N₂
MDL Number:MFCD29767574