Order Entry
Netherlands
ContactUsLinkComponent
tert-Butyl 4-[6-chloro-5-(difluoromethyl)pyrimidin-4-yl]piperazine-1-carboxylate
tert-Butyl 4-[6-chloro-5-(difluoromethyl)pyrimidin-4-yl]piperazine-1-carboxylate
Catalog # APOSPC520062-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 4-[6-chloro-5-(difluoromethyl)pyrimidin-4-yl]piperazine-1-carboxylate
Catalog # APOSPC520062-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :348.776 g/mol
Storage Temperature:Ambient
CAS Number:2149590-34-9
Formula:C₁₄H₁₉ClF₂N₄O₂
MDL Number:MFCD29767545