tert-Butyl N-[6-chloro-4-(difluoromethyl)-3-pyridyl]carbamate
Catalog # APOSPC520042-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:278.683 g/mol
Storage Temperature:Ambient
CAS Number:2149597-34-0
Formula:C₁₁H₁₃ClF₂N₂O₂
MDL Number:MFCD29767525