tert-Butyl N-[3-(difluoromethyl)-2-pyridyl]carbamate
Catalog # APOSPC520039-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:244.238 g/mol
Storage Temperature:Ambient
CAS Number:1816284-42-0
Formula:C₁₁H₁₄F₂N₂O₂
MDL Number:MFCD29767523