Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-[6-(difluoromethyl)-2-pyridyl]benzoate
Methyl 3-[6-(difluoromethyl)-2-pyridyl]benzoate
Methyl 3-[6-(difluoromethyl)-2-pyridyl]benzoate
APOSPC520026-1G
:
Apollo Scientific
:
Methyl 3-[6-(difluoromethyl)-2-pyridyl]benzoate
APOSPC520026-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
263.239 g/mol
:
Ambient
:
C₁₄H₁₁F₂NO₂
:
MFCD29767510