Order Entry
Netherlands
ContactUsLinkComponent
 
4-[6-(Difluoromethyl)-2-pyridyl]aniline
Catalog # APOSPC520020-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[6-(Difluoromethyl)-2-pyridyl]aniline
Catalog # APOSPC520020-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.218 g/mol
Storage Temperature:Ambient
CAS Number:2092578-02-2
Formula:C₁₂H₁₀F₂N₂
MDL Number:MFCD29767505