tert-Butyl N-[4-[6-(difluoromethyl)-2-pyridyl]phenyl]carbamate
Catalog # APOSPC520019-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:320.334 g/mol
Storage Temperature:Ambient
Formula:C₁₇H₁₈F₂N₂O₂
MDL Number:MFCD29767504