
Methyl 2,6-difluoro-4-(trifluoromethoxy)benzoate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2267343-62-2
:
Formula:C₉H₅F₅O₃
MDL Number:MFCD32219445
:256.126 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together