
4-(Aminomethyl)-2-(difluoromethyl)pyridine, 4-BOC protected 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1428532-94-8
:
Formula:C₁₂H₁₆F₂N₂O₂
MDL Number:MFCD23701108
:258.264 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together