4-Chloro-2-(trichloromethyl)-8-(trifluoromethyl)quinoline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:91991-76-3
:
Formula:C₁₁H₄Cl₄F₃N
MDL Number:MFCD02179716
:348.963 g/mol
Storage Temperature:Ambient
Frequently Bought Together