4-(4-Fluorophenyl)-1-(p-tolyl)-1-butanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2006277-31-0
:
Formula:C₁₇H₁₇FO
MDL Number:MFCD29049235
:256.315 g/mol
Storage Temperature:Kamertemperatuur
Frequently Bought Together