Methyl 3-Fluoro-4-(4-methyl-1-piperazinyl)benzoate
Catalog # APOSPC510084-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:252.285 g/mol
Storage Temperature:Ambient
CAS Number:948018-58-4
Formula:C₁₃H₁₇FN₂O₂
MDL Number:MFCD20134210