[6-[3-(Trifluoromethyl)phenyl]-3-pyridyl]methanol
Catalog # APOSPC510076-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Verpakking:Bottle
- VPE:5 g
- Seq:3173
Specifications
About this item
:
:253.22 g/mol
Storage Temperature:Kamertemperatuur
CAS Number:887974-41-6
Formula:C₁₃H₁₀F₃NO
MDL Number:MFCD06410447