[6-[4-(Trifluoromethoxy)phenyl]-3-pyridyl]methanol
Catalog # APOSPC510072-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:269.219 g/mol
Storage Temperature:Ambient
CAS Number:851069-96-0
Formula:C₁₃H₁₀F₃NO₂
MDL Number:MFCD06410451