5-[3-(Trifluoromethoxy)phenyl]nicotinic acid
Catalog # APOSPC510046-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:283.203 g/mol
Storage Temperature:Ambient
CAS Number:924854-23-9
Formula:C₁₃H₈F₃NO₃
MDL Number:MFCD06410278