Methyl 5-Fluoro-2-(4-methylpiperazino)benzoate
Catalog # APOSPC510021-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Verpakking:Bottle
- VPE:5 g
- Seq:3173
Specifications
About this item
:
:252.285 g/mol
Storage Temperature:Kamertemperatuur
CAS Number:1256633-14-3
Formula:C₁₃H₁₇FN₂O₂
MDL Number:MFCD18071007