2-(5,7-Difluoro-6-quinolyl)propanoic acid
Catalog # APOSPC510018-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:237.202 g/mol
Storage Temperature:Ambient
CAS Number:1226776-94-8
Formula:C₁₂H₉F₂NO₂
MDL Number:MFCD16495907