
1-[2-Bromo-4-(trifluoromethyl)phenyl]-3-(2-chloroethyl)urea
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2407339-55-1
:
Formula:C₁₀H₉BrClF₃N₂O
MDL Number:MFCD32264076
:345.543 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together