
1-Chloro-2-fluoro-4-methoxy-5-nitrobenzene 97%
Catalog # APOSPC50377-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.571 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1089280-66-9
Formula:C₇H₅ClFNO₃
MDL Number:MFCD11847613