
Methyl 2-bromo-4-chloro-6-fluorobenzoate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:943975-33-5
:
Formula:C₈H₅BrClFO₂
MDL Number:MFCD28733924
:267.48 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together