4-Amino-2,6-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502875-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:325.182 g/mol
Storage Temperature:Ambient
CAS Number:2244088-51-3
Formula:C₁₃H₆F₇NO
MDL Number:MFCD31630100