2-Amino-4-bromo-5-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502869-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:386.087 g/mol
Storage Temperature:Ambient
CAS Number:2244088-78-4
Formula:C₁₃H₆BrF₆NO
MDL Number:MFCD31630082