2-Amino-4-fluoro-5-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502864-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:321.218 g/mol
Storage Temperature:Ambient
CAS Number:2244087-74-7
Formula:C₁₄H₉F₆NO
MDL Number:MFCD31630067