2-Amino-5-bromophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502862-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:368.097 g/mol
Storage Temperature:Ambient
CAS Number:2244087-59-8
Formula:C₁₃H₇BrF₅NO
MDL Number:MFCD31630061