Order Entry
Netherlands
ContactUsLinkComponent
2-Amino-3-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-3-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502859-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Amino-3-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502859-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :303.227 g/mol
Storage Temperature:Ambient
CAS Number:2244086-02-8
Formula:C₁₄H₁₀F₅NO
MDL Number:MFCD31630052