Order Entry
Netherlands
ContactUsLinkComponent
5-Amino-2-chloro-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Amino-2-chloro-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502843-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Amino-2-chloro-4-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502843-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :341.636 g/mol
Storage Temperature:Ambient
CAS Number:2244086-77-7
Formula:C₁₃H₆ClF₆NO
MDL Number:MFCD31630004