Order Entry
Netherlands
ContactUsLinkComponent
 
4-Aminophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502840-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Aminophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502840-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :289.201 g/mol
Storage Temperature:Ambient
CAS Number:2244083-67-6
Formula:C₁₃H₈F₅NO
MDL Number:MFCD31629995