Order Entry
Netherlands
ContactUsLinkComponent
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-nitrophenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-nitrophenyl ether 95%
Catalog # APOSPC502833-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-nitrophenyl ether 95%
Catalog # APOSPC502833-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :319.184 g/mol
Storage Temperature:Ambient
CAS Number:2244083-86-9
Formula:C₁₃H₆F₅NO₃
MDL Number:MFCD31629974