5-Chloro-2-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502824-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:338.657 g/mol
Storage Temperature:Ambient
CAS Number:2244086-53-9
Formula:C₁₄H₈ClF₅O₂
MDL Number:MFCD31630096