Order Entry
Netherlands
ContactUsLinkComponent
2-Chloro-4-fluoro-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Chloro-4-fluoro-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502819-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-4-fluoro-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502819-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :452.518 g/mol
Storage Temperature:Ambient
CAS Number:2244084-74-8
Formula:C₁₃H₄ClF₆IO
MDL Number:MFCD31630081