Order Entry
Netherlands
ContactUsLinkComponent
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-5-(trifluoromethyl)phenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-5-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502817-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-5-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502817-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :360.175 g/mol
Storage Temperature:Ambient
CAS Number:2244086-88-0
Formula:C₁₄H₅F₉O
MDL Number:MFCD31630075