Order Entry
Netherlands
ContactUsLinkComponent
3-Chloro-2-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
3-Chloro-2-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502814-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-2-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502814-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :326.622 g/mol
Storage Temperature:Ambient
CAS Number:2244086-21-1
Formula:C₁₃H₅ClF₆O
MDL Number:MFCD31630066