Order Entry
Netherlands
ContactUsLinkComponent
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-methylphenyl ether 95%
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-methylphenyl ether 95%
Catalog # APOSPC502812-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Difluoro-4-(trifluoromethyl)phenyl 2-fluoro-4-methylphenyl ether 95%
Catalog # APOSPC502812-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :306.203 g/mol
Storage Temperature:Ambient
CAS Number:2244084-06-6
Formula:C₁₄H₈F₆O
MDL Number:MFCD31630060