Order Entry
Netherlands
ContactUsLinkComponent
4-Chloro-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Chloro-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502806-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-3-fluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502806-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :326.622 g/mol
Storage Temperature:Ambient
CAS Number:2244086-28-8
Formula:C₁₃H₅ClF₆O
MDL Number:MFCD31630042