Order Entry
Netherlands
ContactUsLinkComponent
5-Bromo-4-fluoro-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
5-Bromo-4-fluoro-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502794-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Bromo-4-fluoro-2-methylphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502794-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :385.099 g/mol
Storage Temperature:Ambient
CAS Number:2244086-89-1
Formula:C₁₄H₇BrF₆O
MDL Number:MFCD31630006