Order Entry
Netherlands
ContactUsLinkComponent
2-Bromo-3-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-3-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502790-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-3-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502790-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :478.979 g/mol
Storage Temperature:Ambient
CAS Number:2244088-26-2
Formula:C₁₃H₅BrF₅IO
MDL Number:MFCD31629994