Order Entry
Netherlands
ContactUsLinkComponent
2-Bromo-4-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Bromo-4-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502789-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-4-(trifluoromethoxy)phenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502789-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :437.08 g/mol
Storage Temperature:Ambient
CAS Number:2244087-03-2
Formula:C₁₄H₅BrF₈O₂
MDL Number:MFCD31629991