Order Entry
Netherlands
ContactUsLinkComponent
4-Bromo-3-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Bromo-3-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502782-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-3-methoxyphenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502782-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :383.108 g/mol
Storage Temperature:Ambient
CAS Number:2244086-35-7
Formula:C₁₄H₈BrF₅O₂
MDL Number:MFCD31629970