4-Bromo-2-chlorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502765-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:387.527 g/mol
Storage Temperature:Ambient
CAS Number:2244084-95-3
Formula:C₁₃H₅BrClF₅O
MDL Number:MFCD31630068