4,5-Dichloro-2-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502764-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:388.074 g/mol
Storage Temperature:Ambient
CAS Number:2244083-95-0
Formula:C₁₃H₄Cl₂F₅NO₃
MDL Number:MFCD31630065