Order Entry
Netherlands
ContactUsLinkComponent
2,4-Difluoro-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2,4-Difluoro-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502746-1G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Difluoro-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502746-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :355.165 g/mol
Storage Temperature:Ambient
CAS Number:2244083-53-0
Formula:C₁₃H₄F₇NO₃
MDL Number:MFCD31630011