2-Chloro-4-fluoro-5-nitrophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Catalog # APOSPC502729-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:371.619 g/mol
Storage Temperature:Ambient
CAS Number:2244085-06-9
Formula:C₁₃H₄ClF₆NO₃
MDL Number:MFCD31629959