
Methyl 2-chloro-6-(trifluoromethoxy)benzoate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1261453-43-3
:
Formula:C₉H₆ClF₃O₃
MDL Number:MFCD18399023
:254.59 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together